Learn More
Medchemexpress LLC Aldosterone | 52-39-1 | 99.6% | 360.44 | 1 MG
SDP
Catalog No. 5000437041
Encompass_Preferred
Aldosterone is the primary mineralocorticoid and a steroid hormone. It is synthesized and secreted by the zona glomerulosa (ZG) of the adrenal cortex in response to renin-angiotensin system (RAS) activation or high dietary potassium. Aldosterone's activity depends on its binding and activation of the cytoplasmic/nuclear mineralocorticoid receptor (MR) at a cellular level.
  • Primary mineralocorticoid
  • Steroid hormone
  • Synthesized and secreted by the zona glomerulosa (ZG) of the adrenal cortex
  • Activity depends on binding and activation of the cytoplasmic/nuclear mineralocorticoid receptor (MR)
Catalog No. 50-004-37041 Supplier Medchemexpress LLC Supplier No. HY1133131MG
Aldosterone is the primary mineralocorticoid and a steroid hormone. It is synthesized and secreted by the zona glomerulosa (ZG) of the adrenal cortex in response to renin-angiotensin system (RAS) activation or high dietary potassium. Aldosterone's activity depends on its binding and activation of the cytoplasmic/nuclear mineralocorticoid receptor (MR) at a cellular level.
  • Primary mineralocorticoid
  • Steroid hormone
  • Synthesized and secreted by the zona glomerulosa (ZG) of the adrenal cortex
  • Activity depends on binding and activation of the cytoplasmic/nuclear mineralocorticoid receptor (MR)