Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aldosterone Competitive ELISA Kit

Catalog No. EIAALD
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAALD 96 Tests
EIAALDX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAALD Supplier Invitrogen™ Supplier No. EIAALD
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Aldosterone Competitive ELISA quantitates aldosterone in extracted serum,plasma,urine,dried fecal extracts or cell culture medium.

Principle of the method

The Aldosterone Competitive ELISA Kit research-use-only kit is designed to quantitatively measure aldosterone present in extracted serum and plasma,or in urine,extracted dried fecal samples,and tissue culture media samples. An aldosterone stock solution is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into the provided plate coated with a capture antibody. An aldosterone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to aldosterone to each well. After incubation,the plate is washed and substrate is added. The substrate reacts with the bound aldosterone-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Aldosterone is a hormone produced by the adrenal glands that helps regulate blood pressure through increasing both the release of potassium and the uptake of water and sodium by the kidneys. It is essential for sodium conservation in the kidney, salivary glands, sweat glands and colon. It plays a central role in the regulation of the plasma sodium (Na+), the extracellular potassium (K+) and arterial blood pressure. It does so mainly by acting on the mineralocorticoid receptors in the distal tubules and collecting ducts of the nephron. It influences the reabsorption of sodium and excretion of potassium (from and into the tubular fluids, respectively) of the kidney, thereby indirectly influencing water retention or loss, blood pressure and blood volume. When dysregulated, aldosterone is pathogenic and contributes to the development and progression of cardiovascular and renal disease. Aldosterone has exactly the opposite function of the atrial natriuretic hormone secreted by the heart.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.906-4000 pg/mL
Assay Sensitivity 4.97 pg/mL
Content And Storage Pre-coated plate(s), standard, detector antibody, HRP conjugate, assay buffer, wash buffer, chromogen, stop solution, plate covers, and lot-specific technical data sheet.

Store at –20°C.
Description Aldosterone Competitive ELISA Kit
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Target Species Chemical
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Absorbance Microplate Reader
Interassay CV 19.5%
Intraassay CV 6.9%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases C21H28O5
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 100 μL, Plasma 100 μL, Serum 100 μL, Supernatant 100 μL, Urine 25 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Aldosterone
Test Time 1 hr 20 min
Total Assay Time 16 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.