Learn More
Arbor Assays Aldosterone ELISA Kit (5 Plate)
SDP
Supplier:  Arbor Assays K052H5
Encompass_Preferred
The DetectX Aldosterone ELISA Kits measure Aldosterone present in extracted serum and plasma, or in urine, saliva, extracted dried fecal samples, and tissue culture media samples.
Catalog No. 50-227-8476