Learn More
Medchemexpress LLC Aldoxorubicin hydrochloride | 480998-12-7 | 98.0% | 787.21 g/mol | C37H43ClN4O13 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16261B10MG
Encompass_Preferred
MC-DOXHZN hydrochloride ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of doxorubicin that releases the active anthracycline under acidic conditions. It acts as a DNA topoisomerase II inhibitor and is commonly used as a research reagent and as an intermediate for constructing drug-antibody conjugates.
  • Albumin-binding prodrug with acid-sensitive release
  • Functions as a DNA topoisomerase II inhibitor
  • Suitable as an intermediate for drug-antibody conjugates
  • Supplied as a solid with high stated purity
  • Small pack sizes available for research applications
Catalog No. 50-002-02832