Learn More
Medchemexpress LLC Aldoxorubicin hydrochloride | 480998-12-7 | 98.0% | 787.21 g/mol | C37H43ClN4O13 | 5 MG
SDP
Catalog No. 5000202833
Encompass_Preferred
Aldoxorubicin hydrochloride is an albumin-binding proagent of doxorubicin that functions as a DNA topoisomerase II inhibitor and exhibits acid-sensitive release properties. It is supplied as a high-purity research reagent for preclinical laboratory use.
  • Albumin-binding proagent of doxorubicin
  • Acid-sensitive release of active doxorubicin
  • Acts as a DNA topoisomerase II inhibitor
  • Available as small-mass research packs (e.g., 5 mg)
  • High reported purity (around 98%)
Catalog No. 50-002-02833 Supplier Medchemexpress LLC Supplier No. HY16261B5MG
Aldoxorubicin hydrochloride is an albumin-binding proagent of doxorubicin that functions as a DNA topoisomerase II inhibitor and exhibits acid-sensitive release properties. It is supplied as a high-purity research reagent for preclinical laboratory use.
  • Albumin-binding proagent of doxorubicin
  • Acid-sensitive release of active doxorubicin
  • Acts as a DNA topoisomerase II inhibitor
  • Available as small-mass research packs (e.g., 5 mg)
  • High reported purity (around 98%)