Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Aldrich 1-Phenylheptane, 98%, Clear Liquid, 25 g

Catalog No. NC4013220 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass

1-Phenylheptane, also known as heptylbenzene, is an organic building block primarily utilized in chemical synthesis and as a stationary phase component in high-temperature liquid chromatography. This compound is capable of forming charge-transfer complexes with materials such as fluoranil and 1,3,5-trinitrobenzene.

  • Assay: 98%
  • Molecular weight: 176.30 g/mol
  • Density: 0.86 g/cm3
  • Boiling point: 233 °C
  • Melting point: -48 °C
  • Refractive index: 1.485 n20/D
  • Physical state: Clear, colourless liquid
  • Applications: Used in zirconia and titania based stationary phase capillary HTLC to separate alkyl benzene derivatives

Catalog No. NC4013220 Supplier Sigma Organic Chemistry Supplier No. 11318225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenylheptane, also known as heptylbenzene, is an organic building block primarily utilized in chemical synthesis and as a stationary phase component in high-temperature liquid chromatography. This compound is capable of forming charge-transfer complexes with materials such as fluoranil and 1,3,5-trinitrobenzene.

  • Assay: 98%
  • Molecular weight: 176.30 g/mol
  • Density: 0.86 g/cm3
  • Boiling point: 233 °C
  • Melting point: -48 °C
  • Refractive index: 1.485 n20/D
  • Physical state: Clear, colourless liquid
  • Applications: Used in zirconia and titania based stationary phase capillary HTLC to separate alkyl benzene derivatives

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.