Learn More
Sigma Aldrich Fine Chemicals Biosciences ATMOSBAG 2HAND NS MICR™ | Non-Sterile Gas Sampling Bag | 1 EA
Catalog No. NC2544187
Encompass
The non-sterile ALDRICH ATMOSBAG 2HAND NS MICR is a gas sampling bag for manual collection and storage of atmospheric samples. It allows microbiological analysis of gas samples. As a non-sterile bag, it is not for aseptic or sterile sampling.
Catalog No. NC2544187 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5643971EA
The non-sterile ALDRICH ATMOSBAG 2HAND NS MICR is a gas sampling bag for manual collection and storage of atmospheric samples. It allows microbiological analysis of gas samples. As a non-sterile bag, it is not for aseptic or sterile sampling.