Learn More
MilliporeSigma™ Supelco™ Aldrich™ Chromatography Sprayer
Catalog No. 111009543 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Capacity (Metric):
10 mL
50 mL
125 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-100-9543 125 mL
11-100-9541 10 mL
11-100-9542 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-9543 Supplier MilliporeSigma™ Supelco™ Supplier No. Z5297371EA
Add to Cart
Add to Cart
Provide a fine, uniform spray that is optimized for the development of TLC plates. Also suited for use in electrophoresis.
Adjustable spray pattern using thumb on vent hole. Greaseless, screw-threaded joint will not seize; a simple turn of the threaded cap pulls joint apart safely. Threaded SafetyBarbs™ prevent accidental breakage during tubing installation or removal. Uses low-pressure gas or air.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 125 mL
For Use With (Application) TLC
Product Line Aldrich
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC