Learn More
Sigma Aldrich Fine Chemicals Biosciences Aldrich clamp holder with1EA
Catalog No. 502901718
Encompass
Aldrich clamp holder with1EA Holds rods from 0.089 to 1.6 cm (0.6 in.) diameter.
Catalog No. 50-290-1718 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2436201EA
Aldrich clamp holder with1EA Holds rods from 0.089 to 1.6 cm (0.6 in.) diameter.