Learn More
Sigma Aldrich Fine Chemicals Biosciences Aldrich(R) Liebig condenser Overall H 300 mm, Joint: ST/NS 14/20 | 1EA
Catalog No. NC1843538
Encompass
Aldrich(R) Liebig condenser Overall H 300 mm, Joint: ST/NS 14/20 | 1EA
Catalog No. NC1843538 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5309801EA
Aldrich(R) Liebig condenser Overall H 300 mm, Joint: ST/NS 14/20 | 1EA