Learn More
Sigma Aldrich Aldrich™ Negative Photoresist Kit I
Catalog No. 6548921KT Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 kt
Catalog No. Quantity
65-489-21KT 1 kt
1 options
Catalog No. 65-489-21KT Supplier Sigma Aldrich Supplier No. 6548921KT
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Aldrich(R) Negative Photoresist Kit I consists of materials for photolithography. The photoresist can be spin coated on the glass substrate for the formation of electrode arrays. The kit also provides masking and patterning on the glass surface.Aldrich(R) Negative Photoresist Kit I may be used in the fabrication of microfluidic chips and grating couples.Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Specifications
Quantity 1 kt