Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR antisplash splash1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5492151EA
Encompass
These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.Application for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of bumping. A drain hole at the base of the inner tube allows solventt to drain back into the evaporation flask.
Catalog No. 50-290-1395