Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR AtmosBag two-hand1EA
Catalog No. 502901906
Encompass
The product is packaged in a sterile-pak envelope and processed with ethylene oxide. An indicator sticker on the sterile-pak verifies that the bag has been processed with ethylene oxide.
Catalog No. 50-290-1906 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1183621EA
The product is packaged in a sterile-pak envelope and processed with ethylene oxide. An indicator sticker on the sterile-pak verifies that the bag has been processed with ethylene oxide.