Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR AtmosBag two-hand1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1128281EA
Encompass
for Laboratory Use AtmosBag glove bag is a flexible inflatable polyethylene chamber with built-in gloves that lets you work in a totally isolated and controlled environment. It is an inexpensive portable alternative to the glove box and is well suited to a variety of tasks and materials.for field Sampling AtmosBag slips over the top of cans and drums to blanket with inert gas and isolate vapors and dusts during sampling operations. Minimizes worker exposure and preserves the integrity of the sampled bulk material. AtmosBag serves as a portable laboratory for conducting tests crime scene investigations and other applications where a clean isolated chamber is needed.AtmosBag with Tape-seal closure The opening of AtmosBag is taped and folded over for a gas-tight seal. Each AtmosBag is supplied with a strip of removable sealing tape.AtmosBag with Zipper-lock closure The opening of AtmosBag is closed with a Zipper-lock for a gas-tight seal. The opening is slightly smaller than the width
Catalog No. 50-290-1164