Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR BenchclampTM 2-pr1EA
Catalog No. 502901237
Encompass
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Catalog No. 50-290-1237 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5567181EA
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.