Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR BenchclampTM 3-pr1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5566611EA
Encompass
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient. Round extension arms allow the clamps to be rotated 360. Extension arms allow placement of apparatus at various distances from lab frames. Supplied with vinyl and fiberglass prong covers for high temperature applications.NOTE An additional clamp holder is required to attach clamps to frames or other apparatus.
Catalog No. 50-290-1244