Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR chemical dryer AC1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6835071EA
Encompass
Dries samples using heat vacuum and desiccantPrecise digital control simultaneously displays both set and actual temperatures from ambient to 230C. A built-in thermocouple automatically maintains selected temperature. The oven features a precision machined aluminum heating block with a borosilicate glass sample-viewing port and an integrated safety guard to prevent touching hot parts. The glass drying tube is protected within the enclosed heating block and is safely contained in the event of accidental breakage. The Modular design permits the removal and storage of dried samples in sealed drying tubes. This enables rapid sample processing in succession for servicing multiple users. Order glass drying tube assembly Z222712 separately.SpecificationsAccuracy.nbspTemperature range ambient to 230C.nbspCalibration accuracy 0.1% of span.nbspTemperature stability 0.2F/C rise in ambient maximum.nbspVoltage stability 0.01% of span/% of rated line voltagePower.nbsp50/60 Hz 5%.nbspData retention
Catalog No. 50-290-8546