Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR compact ductless1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6872001EA
Encompass
Portable self-contained compact air cleaning systems capture air contaminants directly at the source and safely remove them before clean air is released into the room. Optional castor base allows easy movement of system to locations throughout the lab. Fume hood shown in image not included.Typical applicationsChemical vapors and dustsCleaning/working with solventsClean room applicationsSoldering and welding operationsFilter SelectionA powerful blower pulls contaminated air from the source through a series of filters and adsorption media to effectively remove particulates and vapors. Various filter media options are available and are matched to the specific contaminants to get the best results. All systems come with the prefilter (Z687383). Systems do not include filter media modules. See the Air Cleaner System Selection Guide for choosing appropriate filter media modules. The HEPA Filter Modules are for applications where there are visible airborne particulates. Th
Catalog No. 50-290-8798