Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR concentrator cond1EA
Catalog No. 502905895
Encompass
Efficiently condenses vapors for rapid solvent removal. Vapors rise up the inner tube and condense on the highly efficient spiral path.features. Replaces traditional 3-piece distillation set-up. Compact size only 30 cm high. Use under vacuum or at atmospheric pressure. Water cooled jacket
Catalog No. 50-290-5895 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4223041EA
Efficiently condenses vapors for rapid solvent removal. Vapors rise up the inner tube and condense on the highly efficient spiral path.features. Replaces traditional 3-piece distillation set-up. Compact size only 30 cm high. Use under vacuum or at atmospheric pressure. Water cooled jacket