Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR corrosive lecture1EA
Catalog No. 502908838
Encompass
Single-stageCGA inlet connection needle valve outlet with A¼ in. NPTM connectionMax. inlet pressure 0-4000 psigTemp. range -40 to 140F316 Stainless Steel body 1 A½ in. gauge face PTFE seals and inner friction sleevesNPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure order bonnet-vent tubing connectors separatelyUse with T-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipmentUse for high-purity applicationsLecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.
Catalog No. 50-290-8838 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5129741EA
Single-stageCGA inlet connection needle valve outlet with A¼ in. NPTM connectionMax. inlet pressure 0-4000 psigTemp. range -40 to 140F316 Stainless Steel body 1 A½ in. gauge face PTFE seals and inner friction sleevesNPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure order bonnet-vent tubing connectors separatelyUse with T-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipmentUse for high-purity applicationsLecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.