Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR dual action NMR t1EA
Catalog No. 502908665
Encompass
Washes tubes inside and out simultaneouslySpecially designed to prevent accidental breakage of tubesOptional 1 000 mL reservoir flasksHighly efficient solvent jets improve cleaning and reduce solvent usageFor 7 or 8 in. L 5 mm NMR tubesPear-shaped filter flask sold separately.
Catalog No. 50-290-8665 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4141741EA
Washes tubes inside and out simultaneouslySpecially designed to prevent accidental breakage of tubesOptional 1 000 mL reservoir flasksHighly efficient solvent jets improve cleaning and reduce solvent usageFor 7 or 8 in. L 5 mm NMR tubesPear-shaped filter flask sold separately.