Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR Essentials beaker1EA
Catalog No. 502908936
Encompass
Borosilicate glass Griffin style low form beakers with reference graduations and pour spout. Uniform standard walls and flat bottom for even heating. Meet ASTM E960 designation.
Catalog No. 50-290-8936 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7405911EA
Borosilicate glass Griffin style low form beakers with reference graduations and pour spout. Uniform standard walls and flat bottom for even heating. Meet ASTM E960 designation.