Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR Essentials membra1EA
Catalog No. 502905690
Encompass
Clear borosilicate glass membrane filtration apparatus for popular 47mm filter paper and nylon membranes for air water and waste water analysis. Complete apparatus includes Stainless steel pinch clamp filter flask funnel and Course porosity (90-150) membrane support piece with ground joint connection. Unit dis-assembles for easy cleaning. Membranes not included.
Catalog No. 50-290-5690 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7410311EA
Clear borosilicate glass membrane filtration apparatus for popular 47mm filter paper and nylon membranes for air water and waste water analysis. Complete apparatus includes Stainless steel pinch clamp filter flask funnel and Course porosity (90-150) membrane support piece with ground joint connection. Unit dis-assembles for easy cleaning. Membranes not included.