Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR Essentials round
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7411204EA
Encompass
Borosilicate glass wide mouth media/reagent bottles with polypropylene GL 45 thread screw cap with pour ring. Large marking spot and reference graduations.
Catalog No. 50-290-1297