Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR firestone valve c1EA
Catalog No. 502905831
Encompass
A rapid efficient and foolproof purge valve for 100% replacement of air in reaction vessels. Automatically controls gas flow and pressure. No special fittings needed. Connect reaction vessel house-vacuum and purge gas to the firestone valve via 10mm O.D. connections. Conserve expensive gases by decreasing flow to almost zero after purging.
Catalog No. 50-290-5831 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1036161EA
A rapid efficient and foolproof purge valve for 100% replacement of air in reaction vessels. Automatically controls gas flow and pressure. No special fittings needed. Connect reaction vessel house-vacuum and purge gas to the firestone valve via 10mm O.D. connections. Conserve expensive gases by decreasing flow to almost zero after purging.