Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR glass reference e1EA
Catalog No. 502905401
Encompass
The reference electrode is used as a pair with an indication electrode to make a measurement. All types supplied with a 30 in. lead. All reference half-cells have a universal reference pin connector.Help in selecting the correct electrode can be viewed at Electrode Selection GuideHints on Electrode Maintenance
Catalog No. 50-290-5401 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1131071EA
The reference electrode is used as a pair with an indication electrode to make a measurement. All types supplied with a 30 in. lead. All reference half-cells have a universal reference pin connector.Help in selecting the correct electrode can be viewed at Electrode Selection GuideHints on Electrode Maintenance