Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR glass reference e1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1130851EA
Encompass
The reference electrode is used as a pair with an indication electrode to make a measurement. All types supplied with a 30 in. lead. All reference half-cells have a universal reference pin connector.Help in selecting the correct electrode can be viewed at Electrode Selection GuideHints on Electrode Maintenance
Catalog No. 50-290-0429