Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR jacketed round bo1EA
Catalog No. 502908652
Encompass
24/40 joint with two hose connections. Multifunctional-can be water cooled or heated with hot water or steam.
Catalog No. 50-290-8652 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1530871EA
24/40 joint with two hose connections. Multifunctional-can be water cooled or heated with hot water or steam.