Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR KugelrohrTM short1EA
Catalog No. 502908600
Encompass
Quickly Distills Difficult Materials with Minimal Hold-up and LossHigh Vacuum Capability and Safe Front Loading DesignDistillation Applications.nbspHeat-sensitive compounds.nbspLiquids and low-melting solids from polymers and tars.nbspRemoves color and particulates.nbspSuitable for sublimation and solvent evaporationFeatures.nbspAir bath oven distills up to 230C.nbsp25 mL to 2 L flask capacity.nbspFront Loading oven door with safety glass viewing window.nbspInterchangeable PTFE support bearings for flask joint 14/20.nbspDigital thermocouple temperature control.nbspFront panel with controls for temperature vacuum and motor drive.nbspAdjustable receiving flask cooling bath.nbspOscillating drive motorSpecifications.nbspOven flask size range 25 mL to 2 L.nbspHeating range ambient to 230C /- 3C across span after stabilization.nbspPower 120/240 VAC /- 10% 50/60 Hz /- 3% detachable power cord.nbspFuses Heater 7.5 A Vac 2.5 A Power input 10 A.nbspHeater 900 W.nbspSize 25 in. W x 11 in.
Catalog No. 50-290-8600 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6834771EA
Add to Cart
Add to Cart
Request a Quote
Quickly Distills Difficult Materials with Minimal Hold-up and LossHigh Vacuum Capability and Safe Front Loading DesignDistillation Applications.nbspHeat-sensitive compounds.nbspLiquids and low-melting solids from polymers and tars.nbspRemoves color and particulates.nbspSuitable for sublimation and solvent evaporationFeatures.nbspAir bath oven distills up to 230C.nbsp25 mL to 2 L flask capacity.nbspFront Loading oven door with safety glass viewing window.nbspInterchangeable PTFE support bearings for flask joint 14/20.nbspDigital thermocouple temperature control.nbspFront panel with controls for temperature vacuum and motor drive.nbspAdjustable receiving flask cooling bath.nbspOscillating drive motorSpecifications.nbspOven flask size range 25 mL to 2 L.nbspHeating range ambient to 230C /- 3C across span after stabilization.nbspPower 120/240 VAC /- 10% 50/60 Hz /- 3% detachable power cord.nbspFuses Heater 7.5 A Vac 2.5 A Power input 10 A.nbspHeater 900 W.nbspSize 25 in. W x 11 in.