Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR large capacity co1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1031011EA
Encompass
With removable cold-fingers and a choice of 55/50 joint or 50Amm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Catalog No. 50-290-8996