Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR large capacity co1EA
Catalog No. 502908715
Encompass
With removable cold-fingers and a choice of 55/50 joint or 50Amm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Catalog No. 50-290-8715 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2845051EA
With removable cold-fingers and a choice of 55/50 joint or 50Amm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.