Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR large capacity fr1EA
Catalog No. 502908565
Encompass
Filter large volumes of air and moisture-sensitive compounds quickly. With porosity C frit 75 mm O-ring top and 24/40 or 29/32 bottom joint. Choose the appropriate 75 mm O-ring top/joint size to fit. The O-ring top and Stainless steel clamps are sold separately.
Catalog No. 50-290-8565 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5172911EA
Filter large volumes of air and moisture-sensitive compounds quickly. With porosity C frit 75 mm O-ring top and 24/40 or 29/32 bottom joint. Choose the appropriate 75 mm O-ring top/joint size to fit. The O-ring top and Stainless steel clamps are sold separately.