Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR lecture bottle st1EA
Catalog No. 502909030
Encompass
For inert and non-corrosive gas applications.Regulator specifications Single stage panel-mounted brass body with stainless steel pressure diaphragm brass nozzle polyurethane seat nylon seals self seating type relief valve 11/2 in. diameter gauges 3000 psig maximum inlet pressure 0-80 psig delivery range brass outlet needle valve with 1/4 in. NPTF connection.
Catalog No. 50-290-9030 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5625991EA
For inert and non-corrosive gas applications.Regulator specifications Single stage panel-mounted brass body with stainless steel pressure diaphragm brass nozzle polyurethane seat nylon seals self seating type relief valve 11/2 in. diameter gauges 3000 psig maximum inlet pressure 0-80 psig delivery range brass outlet needle valve with 1/4 in. NPTF connection.