Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR low form dewar fl1EA
Catalog No. 502905131
Encompass
for use in reactions requiring low temperatures and magnetic stirring. Shallow design in hemispherical or cylindrical (semi-flat bottom) configurations. Borosilicate glass with protective outer aluminum housing.
Catalog No. 50-290-5131 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2026571EA
for use in reactions requiring low temperatures and magnetic stirring. Shallow design in hemispherical or cylindrical (semi-flat bottom) configurations. Borosilicate glass with protective outer aluminum housing.