Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR macro micro KBr p1EA
Catalog No. 502908966
Encompass
For producing 13mm diameter KBr pellets. Use in conjunction with a 12 ton lab press and a vacuum pump capable of reducing pressure to 2mm to reduce moisture in the sample. Heavy paper inserts in a variety of smaller apertures are available separately for making micropellets with the 13mm die set and also provide a useful frame for mounting and storing the pellets.Hardened stainless steel piston and lower sectionBlack anodized aluminum bodyOptically polished anvilsViton(R) sealsEvacuable via 1/4in. O.D. hose connection
Catalog No. 50-290-8966 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5066991EA
For producing 13mm diameter KBr pellets. Use in conjunction with a 12 ton lab press and a vacuum pump capable of reducing pressure to 2mm to reduce moisture in the sample. Heavy paper inserts in a variety of smaller apertures are available separately for making micropellets with the 13mm die set and also provide a useful frame for mounting and storing the pellets.Hardened stainless steel piston and lower sectionBlack anodized aluminum bodyOptically polished anvilsViton(R) sealsEvacuable via 1/4in. O.D. hose connection