Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR Micro Photochemic1EA
Catalog No. 502905726
Encompass
This newly designed apparatus enables researchers to process up to 16 samples at once. Safe LED light provides the energy needed for the photoreactions.
Catalog No. 50-290-5726 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ALDKIT0011EA
This newly designed apparatus enables researchers to process up to 16 samples at once. Safe LED light provides the energy needed for the photoreactions.