Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR microscale glassw1EA
Catalog No. 502908498
Encompass
High-quality low cost glassware kits designed to cover a broad range of organic reactions in student labs and chemical research. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory. 14/10 ground-glass joints with holed-caps and O-ring seals provide secure gas-tight connections. Economy kit - supplied in a compartmentalized PP storage box with hinged lid. Will do 95% of experiments described in the textbook.Deluxe kit - supplied in a foam-lined plastic carrying case. Covers all experiments in the textbook.
Catalog No. 50-290-8498 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5585671EA
High-quality low cost glassware kits designed to cover a broad range of organic reactions in student labs and chemical research. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory. 14/10 ground-glass joints with holed-caps and O-ring seals provide secure gas-tight connections. Economy kit - supplied in a compartmentalized PP storage box with hinged lid. Will do 95% of experiments described in the textbook.Deluxe kit - supplied in a foam-lined plastic carrying case. Covers all experiments in the textbook.