Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR microscale vial c1EA
Catalog No. 502900613
Encompass
These heavy wall vials have an internal conical bottom and an outside flat ground bottom. The 3-8mL vials have approximate graduations with subdivisions of 1 mL. Includes 20-400 open top nylon screw cap size 014 Viton(R) O-ring and 20mm PTfE-faced silicone septum.
Catalog No. 50-290-0613 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6844301EA
These heavy wall vials have an internal conical bottom and an outside flat ground bottom. The 3-8mL vials have approximate graduations with subdivisions of 1 mL. Includes 20-400 open top nylon screw cap size 014 Viton(R) O-ring and 20mm PTfE-faced silicone septum.