Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR mini gas regulato1EA
Catalog No. 502909045
Encompass
These high-purity single-stage single-gauge regulators have a 316 SS diaphragm for accurate stable pressure control of Aldrich liquefied gases. Delivery pressure 1-50Apsi 1/4 in. NPTF inlet hose barb outlet.
Catalog No. 50-290-9045 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5135471EA
These high-purity single-stage single-gauge regulators have a 316 SS diaphragm for accurate stable pressure control of Aldrich liquefied gases. Delivery pressure 1-50Apsi 1/4 in. NPTF inlet hose barb outlet.