Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR non corrosive lec1EA
Catalog No. 502905634
Encompass
Single-stageInlet CGA 180Outlet male 1/4 NPTfMax. inlet pressure 3 000 psigTemp. range -40 to 140fCompact chrome-plated brass bodyLecture-bottle gas regulators include a PTfE sealing washerOrder spare PTfE sealing washers separately
Catalog No. 50-290-5634 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5129581EA
Single-stageInlet CGA 180Outlet male 1/4 NPTfMax. inlet pressure 3 000 psigTemp. range -40 to 140fCompact chrome-plated brass bodyLecture-bottle gas regulators include a PTfE sealing washerOrder spare PTfE sealing washers separately