Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR student glass lab
Catalog No. 502908986
Encompass
Glassware with standard 14/20 ground-glass joints provides a high-quality low-cost SOLUTIONon for school laboratories and research. Customize the basic 14/20 kit with your choice of glass components and accessories available. Supplied in a foam-lined storage case. Contact our Glass Center to discuss your custom kit requirements at aldglasssial.com
Catalog No. 50-290-8986 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1225481KT
Glassware with standard 14/20 ground-glass joints provides a high-quality low-cost SOLUTIONon for school laboratories and research. Customize the basic 14/20 kit with your choice of glass components and accessories available. Supplied in a foam-lined storage case. Contact our Glass Center to discuss your custom kit requirements at aldglasssial.com