Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR super condenser O1EA
Catalog No. 502908633
Encompass
Extremely efficient condenser design combines a cold-finger coiled Liebig and Vigreux condenser into one unit with a forced spiral path and a very large water-cooled surface area.
Catalog No. 50-290-8633 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2249791EA
Extremely efficient condenser design combines a cold-finger coiled Liebig and Vigreux condenser into one unit with a forced spiral path and a very large water-cooled surface area.