Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR Sure/PacTM statio1EA
Catalog No. 502908594
Encompass
The Sure/Pac station is suitable for inert and non-corrosive liquefied gas applications. Designed to keep Sure/Pac cylinders upright and eliminates tip over. A stable base secures the gas cylinder regulator and connection hose yet remains portable. The system comes completely assembled and ready to use. Table top or wall mountable. Optional Cool/Heat coil warms gas cylinder to maintain gas flow during dispensing.Please Note Only the 275 mL and 900 mL Sure/Pac cylinders will fit the station.SpecificationsSingle stage panel-mounted brass body regulator with stainless steel pressure diaphragm 13/4 inch diameter gauge 300 psig maximum inlet pressure 0 to 15 psig delivery pressure PTFE- lined braided stainless steel extension hose brass swivel fitting and adapter for 1/4 inch NPTM Sure/Pac valve threads. Needle valve outlet with 1/8 in. NPTM connection and 1/8 in. NPTF to 1/4 in. hose barb fitting included.
Catalog No. 50-290-8594 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5664461EA
The Sure/Pac station is suitable for inert and non-corrosive liquefied gas applications. Designed to keep Sure/Pac cylinders upright and eliminates tip over. A stable base secures the gas cylinder regulator and connection hose yet remains portable. The system comes completely assembled and ready to use. Table top or wall mountable. Optional Cool/Heat coil warms gas cylinder to maintain gas flow during dispensing.Please Note Only the 275 mL and 900 mL Sure/Pac cylinders will fit the station.SpecificationsSingle stage panel-mounted brass body regulator with stainless steel pressure diaphragm 13/4 inch diameter gauge 300 psig maximum inlet pressure 0 to 15 psig delivery pressure PTFE- lined braided stainless steel extension hose brass swivel fitting and adapter for 1/4 inch NPTM Sure/Pac valve threads. Needle valve outlet with 1/8 in. NPTM connection and 1/8 in. NPTF to 1/4 in. hose barb fitting included.