Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR two-neck round-bo1EA
Catalog No. 502901099
Encompass
Angled ST joints. Z516929 and Z516953 have 24/40 side joints.
Catalog No. 50-290-1099 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5168721EA
Angled ST joints. Z516929 and Z516953 have 24/40 side joints.