Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR Vigreux column Va1EA
Catalog No. 502905574
Encompass
Vigreux indents run the entire length of the column and act as the packing for the column.Jacketed version is pictured.
Catalog No. 50-290-5574 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4132161EA
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vigreux indents run the entire length of the column and act as the packing for the column.Jacketed version is pictured.