Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR wide mouth dewar1EA
Catalog No. 502905071
Encompass
Borosilicate glass with protective aluminum or plastic mesh housing and extended aluminum base weighted for stability.
Catalog No. 50-290-5071 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1207661EA
Borosilicate glass with protective aluminum or plastic mesh housing and extended aluminum base weighted for stability.