Learn More
TARGETMOL CHEMICALS INC ALE-0540 5MG
SDP
Catalog No. 502579011
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALE-0540 is a nonpeptidic heterocyclic molecule that inhibits the binding of NGF to p75 and TrkA as well as signal transduction and biological responses mediated by TrkA receptors. purity: 97%
Catalog No. 50-257-9011 Supplier TARGETMOL CHEMICALS INC Supplier No. T92925MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALE-0540 is a nonpeptidic heterocyclic molecule that inhibits the binding of NGF to p75 and TrkA as well as signal transduction and biological responses mediated by TrkA receptors. purity: 97%