Learn More
Selleck Chemical LLC Alectinib S2762-50mg
SDP
Catalog No. 503111939
Encompass_Preferred
Alectinib is a potent ALK inhibitor with IC50 of 1 9 nM in cell-free assays sensitive to L1196M mutation and higher selectivity for ALK than PF-02341066 NVP-TAE684 and PHA-E429
Catalog No. 50-311-1939 Supplier Selleck Chemical LLC Supplier No. S276250MG
Alectinib is a potent ALK inhibitor with IC50 of 1 9 nM in cell-free assays sensitive to L1196M mutation and higher selectivity for ALK than PF-02341066 NVP-TAE684 and PHA-E429