Learn More
Selleck Chemical LLC Alectinib (CH5424802) 5mg 1256580-46-7 AF-802, RG-7853
SDP
Supplier:  Selleck Chemical LLC S27625MG
Encompass_Preferred
Alectinib (CH5424802, AF-802, RG-7853) is a potent ALK inhibitor with IC50 of 1.9 nM in cell-free assays, sensitive to L1196M mutation and higher selectivity for ALK than PF-02341066, NVP-TAE684 and PHA-E429. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0698821