Learn More
Medchemexpress LLC Alectinib-d6 | 1616374-19-6 | 98.05% | 1 MG
SDP
Catalog No. 5000340064
Encompass_Preferred
Alectinib-d6 is deuterium labeled Alectinib, a potent, selective, and orally available ALK inhibitor. It effectively penetrates the central nervous system (CNS) and inhibits ALK F1174L and ALK R1275Q with high potency.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-40064 Supplier Medchemexpress LLC Supplier No. HY13011S11MG
Alectinib-d6 is deuterium labeled Alectinib, a potent, selective, and orally available ALK inhibitor. It effectively penetrates the central nervous system (CNS) and inhibits ALK F1174L and ALK R1275Q with high potency.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS